Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018358.2 | NP_060828.2 | 5 | 522 | ||
XM_011512959.2 | XP_011511261.1 | 5 | 526 | |||
GenBank mRNA | AK002060.1 | - | 5 | 499 | ||
AK027288.1 | - | 5 | 397 | |||
AK092415.1 | - | 5 | 405 | |||
AK097073.1 | - | 3 | 782 | |||
AK290106.1 | - | 5 | 394 | |||
AK299256.1 | - | 5 | 401 | |||
AK299433.1 | - | 5 | 286 | |||
BC009253.2 | - | 5 | 401 | 3028775 | ||
BC035652.1 | - | 5 | 396 | |||
BC051754.2 | - | 5 | 396 | 6064628 | ||
BC051884.1 | - | 5 | 396 | 5105761 | ||
DB079392.1 | - | 5 | 454 | |||
KU178694.1 | - | 5 | 389 | |||
KU178695.1 | - | 5 | 389 |