Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252148.1 | NP_001239077.1 | 6 | 1706 | ||
NM_001252151.1 | NP_001239080.1 | 7 | 1202 | |||
NM_001252152.1 | NP_001239081.1 | 8 | 1190 | |||
NM_018375.4 | NP_060845.2 | 7 | 1775 | |||
XM_005267834.1 | XP_005267891.1 | 6 | 1577 | |||
XM_017021426.1 | XP_016876915.1 | 8 | 1916 | |||
GenBank mRNA | AK002136.1 | - | 7 | 1751 | ||
AK293222.1 | - | 7 | 1195 | |||
AK311104.1 | - | 2 | 439 | |||
AK316381.1 | - | 8 | 1179 | |||
AY358687.1 | - | 7 | 1221 | |||
BC064383.1 | - | 7 | 1490 | 5737339 |