Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146688.1 | NP_001140160.1 | 11, 12 | 1787 | ||
NM_018433.5 | NP_060903.2 | 11, 12 | 2056 | |||
XM_006712051.3 | XP_006712114.1 | 10, 11 | 1759 | |||
XM_017004493.1 | XP_016859982.1 | 11, 12 | 2075 | |||
XM_017004494.1 | XP_016859983.1 | 12, 13 | 1898 | |||
GenBank mRNA | AB018285.2 | - | 11, 12 | 1819 | ||
AK304276.1 | - | 9, 10 | 1573 | |||
AL832150.1 | - | 11, 12 | 1822 | |||
BC038297.2 | - | 11, 12 | 1833 | |||
BX640698.1 | - | 11, 12 | 1857 | |||
BX647271.1 | - | 11, 12 | 1825 | |||
BX647752.1 | - | 11, 12 | 1885 | |||
BX648841.1 | - | 11, 12 | 1788 | |||
BX649109.1 | - | 11, 12 | 1826 | |||
CR749581.1 | - | 11, 12 | 1824 |