Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF331 SIRNA Assays ›
Gene Symbol:
ZNF331
Gene Name:
zinc finger protein 331
Gene Aliases:
RITA, ZNF361, ZNF463
Chromosome Location:
Chr.19: 53503819 - 53580269 on Build GRCh38
UniGene ID:
Hs.185674
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
109121
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF331
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079906.1
NP_001073375.1
6
1348
NM_001079907.1
NP_001073376.1
6
1368
NM_001253798.1
NP_001240727.1
7
1366
NM_001253799.1
NP_001240728.1
6
1336
NM_001253800.1
NP_001240729.1
6
1295
NM_001253801.2
NP_001240730.1
5
1258
NM_001317113.1
NP_001304042.1
5
1284
NM_001317114.1
NP_001304043.1
6
1201
NM_001317115.1
NP_001304044.1
7
1378
NM_001317116.1
NP_001304045.1
5
1339
NM_001317117.1
NP_001304046.1
6
1288
NM_001317118.1
NP_001304047.1
6
1369
NM_001317119.1
NP_001304048.1
5
1338
NM_001317120.1
NP_001304049.1
7
2322
NM_001317121.1
NP_001304050.1
5
1242
NM_018555.5
NP_061025.5
7
2360
XM_011527076.2
XP_011525378.1
8
2879
XM_011527078.2
XP_011525380.1
8
2478
XM_017026936.1
XP_016882425.1
12
3072
XM_017026937.1
XP_016882426.1
12
2597
XM_017026938.1
XP_016882427.1
7
1591
XM_017026939.1
XP_016882428.1
6
1363
XM_017026940.1
XP_016882429.1
5
1243
GenBank mRNA
AF251515.2
-
6
1302
AF272148.1
-
5
1112
AK075226.1
-
6
1269
AK128736.1
-
7
2246
AK312889.1
-
5
1221
BC009433.1
-
8
1585
3355780
CR749560.1
-
7
2270
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9xzrh:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline