Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018683.3 | NP_061153.1 | 6 | 1099 | ||
GenBank mRNA | AF131742.1 | - | 2 | 459 | ||
AK091976.1 | - | 6 | 1077 | |||
AK297188.1 | - | 5 | 880 | |||
AK301731.1 | - | 5 | 903 | |||
BC013695.1 | - | 6 | 1068 | 3858611 | ||
BC066919.1 | - | 6 | 1077 | 5264729 | ||
BX640603.1 | - | 7 | 1082 |