Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SVOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018711.4 | NP_061181.1 | 16 | 3627 | ||
GenBank mRNA | AL359592.1 | - | 4 | 2045 | ||
BC033587.1 | - | 16 | 3427 | 5243017 | ||
BC094722.1 | - | 16 | 3273 | 4817829 |