Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018713.2 | NP_061183.2 | 4 | 1641 | ||
NR_046437.1 | 4 | 1679 | ||||
XM_006711437.3 | XP_006711500.2 | 4 | 1977 | |||
XM_017001684.1 | XP_016857173.1 | 4 | 1228 | |||
GenBank mRNA | AK090806.1 | - | 4 | 1679 | ||
AL359609.1 | - | 2 | 1248 | |||
AY212919.1 | - | 4 | 1625 | |||
BC036078.1 | - | 4 | 1201 |