Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127370.2 | NP_001120842.1 | 9 | 1352 | ||
NM_001127371.2 | NP_001120843.1 | 7 | 1160 | |||
NM_018719.4 | NP_061189.2 | 8 | 1298 | |||
GenBank mRNA | AK022955.1 | - | 8 | 1272 | ||
AK095965.1 | - | 9 | 1306 | |||
AK292490.1 | - | 8 | 1273 | |||
AK300781.1 | - | 7 | 1120 | |||
AL359619.1 | - | 3 | 424 | |||
AY161168.1 | - | 8 | 1175 | |||
AY161169.1 | - | 8 | 1178 | |||
BC009352.2 | - | 8 | 1190 | 4139111 | ||
BC014630.2 | - | 7 | 1142 | |||
BC025242.1 | - | 8 | 1252 | 4842169 | ||
BC032576.1 | - | 8 | 1263 | 5491780 | ||
BC039823.1 | - | 8 | 1262 | 6057601 |