Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242628.1 | NP_001229557.1 | 2 | 1490 | ||
NM_001242630.1 | NP_001229559.1 | 2 | 1545 | |||
NM_018988.3 | NP_061861.1 | 2 | 2317 | |||
XM_011514699.2 | XP_011513001.1 | 2 | 1506 | |||
GenBank mRNA | AK000337.1 | - | 2 | 2235 | ||
AK027010.1 | - | 2 | 1545 | |||
AK055131.1 | - | 2 | 1459 | |||
BC119005.1 | - | 2 | 1657 | 40085551 | ||
BC124557.1 | - | 2 | 1657 | 40085552 | ||
BC148295.2 | - | 2 | 1664 | 40122213 |