Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019112.3 | NP_061985.2 | 47 | 6726 | ||
XM_006722616.1 | XP_006722679.1 | 47 | 6659 | |||
XM_006722618.2 | XP_006722681.1 | 32 | 4263 | |||
XM_011527628.2 | XP_011525930.1 | 47 | 6587 | |||
XM_011527629.1 | XP_011525931.1 | 47 | 6686 | |||
XM_011527630.1 | XP_011525932.1 | 46 | 6584 | |||
XM_011527631.1 | XP_011525933.1 | 44 | 6266 | |||
XM_011527632.1 | XP_011525934.1 | 43 | 6165 | |||
XM_011527636.2 | XP_011525938.1 | 32 | 4261 | |||
XM_017026142.1 | XP_016881631.1 | 46 | 6458 | |||
GenBank mRNA | AF250238.1 | - | 46 | 6495 | ||
AF328787.1 | - | 46 | 6611 | |||
AK097344.1 | - | 14 | 1789 | |||
AL137481.1 | - | 10 | 1376 |