Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019849.2 | NP_062823.1 | 7 | 1140 | ||
XM_006723284.2 | XP_006723347.1 | 7 | 1012 | |||
XM_011527119.1 | XP_011525421.1 | 6 | 1097 | |||
XM_011527120.1 | XP_011525422.1 | 5 | 1005 | |||
GenBank mRNA | AB037670.1 | - | 7 | 1092 | ||
AJ277731.1 | - | 7 | 1101 | |||
AK000846.1 | - | 1 | 454 | |||
AK316594.1 | - | 7 | 1012 | |||
BC035627.1 | - | 7 | 1083 | 5200601 |