Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019849.2 | NP_062823.1 | 11 | 1782 | ||
XM_006723284.2 | XP_006723347.1 | 10 | 1417 | |||
XM_011527119.1 | XP_011525421.1 | 9 | 2065 | |||
XM_011527120.1 | XP_011525422.1 | 8 | 1973 | |||
GenBank mRNA | AB037670.1 | - | 11 | 1734 | ||
AJ277731.1 | - | 11 | 1743 | |||
AK000846.1 | - | 4 | 1422 | |||
BC035627.1 | - | 11 | 1725 | 5200601 |
Gene Symbol: LRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU739456.1 | - | 1 | 56 |