Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAMF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020125.2 | NP_064510.1 | 5 | 1766 | ||
XM_011509777.2 | XP_011508079.1 | 5 | 2879 | |||
XM_011509780.2 | XP_011508082.2 | 4 | 1373 | |||
XM_017001844.1 | XP_016857333.1 | 6 | 2795 | |||
XM_017001846.1 | XP_016857335.1 | 5 | 1653 | |||
GenBank mRNA | AF144235.1 | - | 2 | 847 | ||
AF146761.1 | - | 5 | 1644 | |||
AK074669.1 | - | 5 | 1705 |