Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020208.3 | NP_064593.1 | 11 | 2520 | ||
NM_022405.3 | NP_071800.1 | 10 | 2409 | |||
XM_011533847.2 | XP_011532149.1 | 10 | 2834 | |||
GenBank mRNA | AJ276207.1 | - | 11 | 2447 | ||
AJ276208.1 | - | 10 | 2336 | |||
BC126197.1 | - | 11 | 2460 | 8991913 | ||
BC136431.1 | - | 11 | 2460 | 9020420 | ||
BC144087.1 | - | 11 | 1784 | 9052607 |