Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002033.3 | NP_002024.1 | 1 | 2203 | ||
GenBank mRNA | AF305083.1 | - | 1 | 330 | ||
BC136373.1 | - | 1 | 2083 | 9020360 | ||
BC136374.1 | - | 1 | 2083 | 9020361 | ||
M58596.1 | - | 1 | 1602 | |||
M58597.1 | - | 1 | 2093 |