Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBXIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317734.1 | NP_001304663.1 | 9 | 1220 | ||
NM_001317735.1 | NP_001304664.1 | 7 | 889 | |||
NM_020524.3 | NP_065385.2 | 10 | 1309 | |||
GenBank mRNA | AF221521.1 | - | 10 | 1258 | ||
AK022497.1 | - | 9 | 1179 | |||
AK023219.1 | - | 10 | 1250 | |||
AK300987.1 | - | 6 | 598 | |||
AK303413.1 | - | 7 | 833 | |||
AK303863.1 | - | 7 | 734 | |||
AK315551.1 | - | 10 | 1290 | |||
BC016852.1 | - | 10 | 1237 | 3900461 |