Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SORCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020777.2 | NP_065828.2 | 23 | 3097 | ||
XM_005247987.4 | XP_005248044.2 | 23 | 3137 | |||
XM_011513514.2 | XP_011511816.1 | 23 | 3137 | |||
XM_011513515.2 | XP_011511817.1 | 23 | 3137 | |||
XM_011513516.2 | XP_011511818.1 | 23 | 3137 | |||
XM_017008481.1 | XP_016863970.1 | 23 | 3137 | |||
GenBank mRNA | AB037750.1 | - | 20 | 2341 | ||
AK295629.1 | - | 23 | 2713 |