Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020828.1 | NP_065879.1 | 8 | 3362 | ||
XM_011526462.2 | XP_011524764.1 | 8 | 3358 | |||
XM_011526463.2 | XP_011524765.2 | 8 | 4091 | |||
GenBank mRNA | AB037852.1 | - | 8 | 3362 | ||
AF507045.1 | - | 8 | 3362 | |||
CR749659.1 | - | 2640 |