Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOMEZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020834.2 | NP_065885.2 | 2 | 3437 | ||
GenBank mRNA | AF463523.1 | - | 2 | 3358 |