Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324188.1 | NP_001311117.1 | 3 | 2305 | ||
NM_001324189.1 | NP_001311118.1 | 3 | 2228 | |||
NM_020873.6 | NP_065924.3 | 2 | 2023 | |||
GenBank mRNA | AB040930.1 | - | 2 | 4522 | ||
AB074161.1 | - | 1 | 1256 | |||
AK023273.1 | - | 1 | 410 | |||
AK291717.1 | - | 2 | 1607 | |||
AY358312.1 | - | 1 | 1370 | |||
BC034947.2 | - | 2 | 1586 | 4826337 |