Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033561.1 | NP_001028733.1 | 4 | 877 | ||
NM_001290131.1 | NP_001277060.1 | 4 | 877 | |||
NM_020889.2 | NP_065940.1 | 4 | 877 | |||
XM_005258015.3 | XP_005258072.1 | 3 | 166 | |||
XM_005258016.3 | XP_005258073.1 | 3 | 186 | |||
XM_011525078.2 | XP_011523380.1 | 3 | 478 | |||
XM_011525079.2 | XP_011523381.1 | 4 | 860 | |||
XM_011525082.2 | XP_011523384.1 | 4 | 860 | |||
XM_011525083.2 | XP_011523385.1 | 4 | 860 | |||
XM_017024903.1 | XP_016880392.1 | 3 | 925 | |||
XM_017024904.1 | XP_016880393.1 | 3 | 445 | |||
GenBank mRNA | AK160370.1 | - | 4 | 2904 | ||
AK291420.1 | - | 4 | 349 | |||
AK294056.1 | - | 3 | 306 | |||
AY030283.1 | - | 4 | 879 | |||
BC001657.1 | - | 2 | 94 | |||
BC110882.1 | - | 4 | 694 | 6157238 | ||
BC121043.1 | - | 4 | 379 | 40117907 | ||
BC121044.1 | - | 4 | 379 | 40117911 |