Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF529 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145649.1 | NP_001139121.1 | 6 | 2916 | ||
NM_001145650.1 | NP_001139122.1 | 5 | 2856 | |||
NM_001321351.1 | NP_001308280.1 | 6 | 2935 | |||
NM_020951.4 | NP_066002.3 | 5 | 2834 | |||
XM_006723302.3 | XP_006723365.1 | 5 | 2708 | |||
XM_011527164.2 | XP_011525466.1 | 6 | 2851 | |||
XM_011527165.2 | XP_011525467.1 | 4 | 2701 | |||
XM_011527166.2 | XP_011525468.1 | 6 | 3915 | |||
XM_011527167.2 | XP_011525469.1 | 7 | 3035 | |||
XM_017027039.1 | XP_016882528.1 | 5 | 2791 | |||
XM_017027040.1 | XP_016882529.1 | 5 | 6483 | |||
XM_017027041.1 | XP_016882530.1 | 6 | 5571 | |||
XM_017027042.1 | XP_016882531.1 | 7 | 3069 | |||
XM_017027043.1 | XP_016882532.1 | 6 | 2934 | |||
XM_017027044.1 | XP_016882533.1 | 6 | 2976 | |||
GenBank mRNA | AB046835.1 | - | 1 | 2399 | ||
AK025110.1 | - | 6 | 2927 | |||
BC036578.2 | - | 1 | 647 | |||
BC064690.1 | - | 5 | 2796 | 30335277 |