Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021074.4 | NP_066552.2 | 8 | 810 | ||
XM_017025782.1 | XP_016881271.1 | 8 | 1000 | |||
GenBank mRNA | AF147367.1 | - | 4 | 285 | ||
AK291865.1 | - | 8 | 706 | |||
AK312129.1 | - | 8 | 728 | |||
BC001632.1 | - | 8 | 720 | 3536345 | ||
BC017487.1 | - | 8 | 718 | 3537815 | ||
BC071689.1 | - | 8 | 700 | 4372795 | ||
BG742948.1 | - | 8 | 810 | |||
CR456928.1 | - | 8 | 696 | |||
M22538.1 | - | 8 | 714 |