Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021083.2 | NP_066569.1 | 3 | 2322 | ||
GenBank mRNA | AY534238.1 | - | 3 | 2446 | ||
BC036019.1 | - | 3 | 2363 | 5272840 | ||
Z32684.2 | - | 3 | 2322 |