Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAMF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282588.1 | NP_001269517.1 | 4 | 894 | ||
NM_001282589.1 | NP_001269518.1 | 5 | 998 | |||
NM_001282590.1 | NP_001269519.1 | 6 | 1070 | |||
NM_001282591.1 | NP_001269520.1 | 5 | 950 | |||
NM_001282592.1 | NP_001269521.1 | 6 | 1287 | |||
NM_001282593.1 | NP_001269522.1 | 4 | 846 | |||
NM_001282594.1 | NP_001269523.1 | 6 | 1109 | |||
NM_001282595.1 | NP_001269524.1 | 6 | 852 | |||
NM_001282596.1 | NP_001269525.1 | 5 | 966 | |||
NM_021181.4 | NP_067004.3 | 7 | 1391 | |||
XM_011509828.1 | XP_011508130.1 | 7 | 1240 | |||
XM_011509829.1 | XP_011508131.1 | 6 | 1136 | |||
GenBank mRNA | AB027233.1 | - | 7 | 1161 | ||
AF390894.1 | - | 7 | 1166 | |||
AK290706.1 | - | 6 | 1071 | |||
AK292097.1 | - | 6 | 854 | |||
AK292148.1 | - | 7 | 1175 | |||
AK298499.1 | - | 5 | 805 | |||
AK298548.1 | - | 4 | 702 | |||
AK301137.1 | - | 4 | 637 | |||
AK301432.1 | - | 6 | 895 | |||
AK301438.1 | - | 5 | 739 | |||
AL713801.1 | - | 2 | 2016 | |||
AL834424.1 | - | 6 | 859 | |||
BC027867.1 | - | 6 | 1053 | 5203638 |