Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021217.2 | NP_067040.1 | 4 | 1728 | ||
XM_017027081.1 | XP_016882570.1 | 3 | 1591 | |||
XM_017027082.1 | XP_016882571.1 | 4 | 1753 | |||
GenBank mRNA | AK290979.1 | - | 4 | 1664 | ||
AL365370.1 | - | 1 | 792 | |||
BC043354.1 | - | 4 | 1689 | 6014754 | ||
BC062561.1 | - | 4 | 1690 | 6644579 |