Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDA2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199687.2 | NP_001186616.1 | 7 | 2229 | ||
NM_001242310.1 | NP_001229239.1 | 7 | 2194 | |||
NM_001324199.1 | NP_001311128.1 | 5 | 1875 | |||
NM_001324201.1 | NP_001311130.1 | 6 | 2149 | |||
NM_001324202.1 | NP_001311131.1 | 5 | 1578 | |||
NM_001324204.1 | NP_001311133.1 | 5 | 1677 | |||
NM_001324205.1 | NP_001311134.1 | 5 | 1671 | |||
NM_001324206.1 | NP_001311135.1 | 7 | 2237 | |||
NM_021783.4 | NP_068555.1 | 7 | 2231 | |||
NR_136726.1 | 7 | 2138 | ||||
NR_136727.1 | 7 | 2140 | ||||
XM_011531000.1 | XP_011529302.1 | 8 | 2244 | |||
XM_011531001.1 | XP_011529303.1 | 8 | 2292 | |||
XM_011531002.1 | XP_011529304.1 | 8 | 1949 | |||
XM_011531003.2 | XP_011529305.1 | 6 | 2044 | |||
XM_011531005.2 | XP_011529307.1 | 5 | 1878 | |||
XM_011531006.2 | XP_011529308.1 | 6 | 1743 | |||
XM_017029703.1 | XP_016885192.1 | 6 | 2141 | |||
XM_017029704.1 | XP_016885193.1 | 7 | 1886 | |||
XM_017029705.1 | XP_016885194.1 | 6 | 2019 | |||
XM_017029706.1 | XP_016885195.1 | 6 | 1794 | |||
XM_017029707.1 | XP_016885196.1 | 6 | 1928 | |||
XM_017029708.1 | XP_016885197.1 | 5 | 1570 | |||
GenBank mRNA | BC034919.1 | - | 7 | 2178 | 5434545 |