Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002080.3 | NP_002071.2 | 3 | 419 | ||
GenBank mRNA | AK098313.1 | - | 3 | 341 | ||
AK223271.1 | - | 3 | 354 | |||
AK290847.1 | - | 3 | 352 | |||
AK302249.1 | - | 3 | 378 | |||
AK307598.1 | - | 3 | 306 | |||
AL516773.3 | - | 3 | 404 | |||
BC000525.2 | - | 3 | 342 | 2961181 | ||
BI667777.1 | - | 3 | 424 | |||
DB485399.1 | - | 3 | 413 | |||
M22632.1 | - | 3 | 298 |