Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021951.2 | NP_068770.2 | 4 | 1023 | ||
XM_006716732.1 | XP_006716795.1 | 4 | 1069 | |||
XM_011517770.1 | XP_011516072.1 | 4 | 962 | |||
XM_011517771.1 | XP_011516073.1 | 4 | 959 | |||
XM_011517773.2 | XP_011516075.1 | 4 | 781 | |||
XM_017014374.1 | XP_016869863.1 | 3 | 675 | |||
XM_017014375.1 | XP_016869864.1 | 3 | 782 | |||
GenBank mRNA | AF130728.1 | - | 4 | 1023 | ||
AJ276801.1 | - | 4 | 932 | |||
AK310336.1 | - | 4 | 761 | |||
AK313594.1 | - | 4 | 1016 | |||
AL162131.1 | - | 4 | 932 | |||
BC040847.1 | - | 4 | 910 | 5168563 |