Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLOC1S5-TXNDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037616.1 | 10 | 1001 | |||
GenBank mRNA | BC052310.1 | - | 10 | 951 | 6671945 |
Gene Symbol: TXNDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145549.2 | NP_001139021.1 | 7 | 844 | ||
NM_030810.3 | NP_110437.2 | 7 | 880 | |||
GenBank mRNA | AJ440721.1 | - | 7 | 635 | ||
AK025006.1 | - | 1 | 620 | |||
AK075291.1 | - | 7 | 854 | |||
AK098089.1 | - | 9 | 941 | |||
AK315598.1 | - | 7 | 845 | |||
AL833008.1 | - | 7 | 724 | |||
AL834423.1 | - | 4 | 278 | |||
AY326464.1 | - | 7 | 868 | |||
AY358646.1 | - | 7 | 874 | |||
BC001199.1 | - | 7 | 599 | 3356029 | ||
BU855393.1 | - | 7 | 661 |