Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145549.2 | NP_001139021.1 | 10 | 2331 | ||
NM_030810.3 | NP_110437.2 | 10 | 2367 | |||
GenBank mRNA | AK025006.1 | - | 4 | 2107 | ||
AK075291.1 | - | 10 | 2322 | |||
AL833008.1 | - | 10 | 2211 | |||
AL834423.1 | - | 7 | 1765 | |||
AY326464.1 | - | 10 | 2373 | |||
AY358646.1 | - | 10 | 2361 | |||
BC001199.1 | - | 10 | 2067 | 3356029 |
Gene Symbol: BLOC1S5-TXNDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037616.1 | 13 | 2488 | |||
GenBank mRNA | BC052310.1 | - | 13 | 2438 | 6671945 |