Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC28A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199633.1 | NP_001186562.1 | 4 | 408 | ||
NM_022127.2 | NP_071410.1 | 5 | 354 | |||
NR_037638.2 | 5 | 584 | ||||
GenBank mRNA | AF305210.1 | - | 5 | 354 | ||
AK292849.1 | - | 4 | 407 | |||
AK304406.1 | - | 5 | 564 | |||
AK310861.1 | - | 5 | 553 | |||
DB218604.1 | - | 4 | 408 |