Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022336.3 | NP_071731.1 | 12 | 1938 | ||
XM_006712204.1 | XP_006712267.1 | 11 | 1918 | |||
XM_011510502.2 | XP_011508804.2 | 11 | 1797 | |||
XM_011510503.2 | XP_011508805.2 | 12 | 1694 | |||
GenBank mRNA | AF130988.1 | - | 12 | 1939 | ||
CR749352.1 | - | 12 | 2047 |