Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC13A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324400.1 | NP_001311329.1 | 18 | 3113 | ||
NM_022444.3 | NP_071889.2 | 15 | 2612 | |||
XM_011516515.2 | XP_011514817.1 | 16 | 2784 | |||
XM_011516516.2 | XP_011514818.1 | 15 | 2682 | |||
XM_011516517.2 | XP_011514819.1 | 17 | 2903 | |||
XM_017012554.1 | XP_016868043.1 | 15 | 2670 | |||
GenBank mRNA | AF260824.1 | - | 15 | 2612 |