Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284306.1 | NP_001271235.1 | 18 | 4021 | ||
NM_001284307.1 | NP_001271236.1 | 20 | 4036 | |||
NM_022473.2 | NP_071918.1 | 19 | 6464 | |||
XM_005254591.2 | XP_005254648.1 | 22 | 6371 | |||
XM_005254592.2 | XP_005254649.1 | 22 | 6110 | |||
XM_017022483.1 | XP_016877972.1 | 21 | 6146 | |||
XM_017022484.1 | XP_016877973.1 | 19 | 5891 | |||
XM_017022485.1 | XP_016877974.1 | 20 | 3770 | |||
GenBank mRNA | AF205632.1 | - | 19 | 6437 | ||
AK024726.1 | - | 8 | 1293 | |||
AK127741.1 | - | 17 | 3003 | |||
AK226132.1 | - | 20 | 3944 | |||
AL831983.1 | - | 18 | 3967 | |||
AL832014.1 | - | 18 | 4245 | |||
AL832352.1 | - | 18 | 4462 | |||
AL832367.1 | - | 17 | 2915 | |||
AL832455.2 | - | 18 | 4377 | |||
AL833301.1 | - | 18 | 4475 | |||
BX648629.1 | - | 18 | 5569 |