Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098477.1 | NP_001091947.1 | 2 | 359 | ||
NM_002092.3 | NP_002083.3 | 2 | 523 | |||
XM_005265681.3 | XP_005265738.1 | 2 | 762 | |||
XM_011531897.2 | XP_011530199.1 | 2 | 188 | |||
GenBank mRNA | AI151431.1 | - | 2 | 359 | ||
AK055499.1 | - | 3 | 299 | |||
AK090755.1 | - | 3 | 303 | |||
AK094806.1 | - | 2 | 328 | |||
AK095799.1 | - | 3 | 289 | |||
AK097055.1 | - | 3 | 369 | |||
AK124124.1 | - | 3 | 440 | |||
AK127161.1 | - | 3 | 398 | |||
AK294612.1 | - | 2 | 223 | |||
AK296287.1 | - | 3 | 358 | |||
AK298883.1 | - | 2 | 181 | |||
AV739732.1 | - | 2 | 275 | |||
DN990192.1 | - | 2 | 531 | |||
U07231.1 | - | 3 | 333 |