Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022568.3 | NP_072090.1 | 4 | 591 | ||
NM_170771.2 | NP_739577.1 | 4 | 591 | |||
GenBank mRNA | AF303134.1 | - | 4 | 582 | ||
AK074266.1 | - | 4 | 584 | |||
AK222848.1 | - | 4 | 584 | |||
AK290784.1 | - | 4 | 538 | |||
BC113862.1 | - | 4 | 561 | 40033493 | ||
BC114473.1 | - | 5 | 599 | 40033488 |