Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135095.1 | NP_001128567.1 | 3 | 269 | ||
NM_022763.3 | NP_073600.3 | 3 | 342 | |||
XM_017007062.1 | XP_016862551.1 | 3 | 322 | |||
XM_017007064.1 | XP_016862553.1 | 3 | 322 | |||
GenBank mRNA | AB098597.1 | - | 2 | 227 | ||
AK127826.1 | - | 3 | 322 | |||
AK223599.1 | - | 3 | 373 | |||
AK314478.1 | - | 3 | 269 | |||
AY358367.1 | - | 3 | 311 | |||
BC012204.1 | - | 3 | 340 | |||
BC039297.1 | - | 3 | 233 | 5297602 | ||
BX648340.1 | - | 3 | 264 |