Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135095.1 | NP_001128567.1 | 23, 24 | 3115 | ||
NM_022763.3 | NP_073600.3 | 23, 24 | 3188 | |||
XM_017007062.1 | XP_016862551.1 | 22, 23 | 2886 | |||
XM_017007063.1 | XP_016862552.1 | 18, 19 | 2372 | |||
GenBank mRNA | AB098597.1 | - | 22, 23 | 3073 | ||
AF543840.1 | - | 5, 6 | 889 | |||
AK027052.1 | - | 7, 8 | 1171 | |||
AK075220.1 | - | 13, 14 | 1810 | |||
AK127826.1 | - | 23, 24 | 3168 | |||
AK223599.1 | - | 23, 24 | 3219 | |||
AK314478.1 | - | 23, 24 | 3115 | |||
AL157482.1 | - | 5, 6 | 836 | |||
AY358146.1 | - | 16, 17 | 2061 | |||
BC033635.2 | - | 11, 12 | 1663 | |||
BC039297.1 | - | 23, 24 | 3079 | 5297602 | ||
BX648340.1 | - | 23, 24 | 3110 |