Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WHSC1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017778.2 | NP_060248.2 | 2 | 692 | ||
NM_023034.1 | NP_075447.1 | 2 | 692 | |||
GenBank mRNA | AF255649.1 | - | 3 | 291 | ||
AF318339.1 | - | 2 | 863 | |||
AF332468.1 | - | 2 | 692 | |||
AF332469.1 | - | 2 | 692 | |||
AJ295990.1 | - | 2 | 487 | |||
AJ295991.1 | - | 2 | 487 | |||
AJ295992.1 | - | 2 | 487 | |||
AK022560.1 | - | 2 | 369 | |||
AK127594.1 | - | 2 | 1401 | |||
BC012059.1 | - | 2 | 660 | |||
BC062631.1 | - | 2 | 631 | |||
BC073858.1 | - | 1 | 218 | |||
BC101717.1 | - | 2 | 266 | 8069223 | ||
BC107734.1 | - | 1 | 668 | |||
BC113469.1 | - | 2 | 266 | 8322521 | ||
BC115006.1 | - | 2 | 287 | 7961962 | ||
BC143510.1 | - | 2 | 266 | 9052018 |