Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYCRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023078.3 | NP_075566.2 | 6 | 1234 | ||
GenBank mRNA | AK001500.2 | - | 6 | 1171 | ||
AK023914.1 | - | 6 | 1194 | |||
AK294615.1 | - | 7 | 1302 | |||
AL833857.1 | - | 5 | 1078 |