Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYCRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023078.3 | NP_075566.2 | 6 | 1869 | ||
GenBank mRNA | AF086378.1 | - | 1 | 219 | ||
AK001500.2 | - | 6 | 1806 | |||
AK023914.1 | - | 6 | 1828 | |||
AK222685.1 | - | 1016 | ||||
AL833857.1 | - | 5 | 1713 |