Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005257286.2 | XP_005257343.1 | 1 | 811 | ||
XM_017024564.1 | XP_016880053.1 | 1 | 811 |
Gene Symbol: HOXB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024017.4 | NP_076922.1 | 2 | 1814 | ||
GenBank mRNA | AK026084.1 | - | 1 | 629 | ||
AK056123.1 | - | 2 | 1593 | |||
AK056414.1 | - | 3 | 1637 | |||
BC015565.1 | - | 3 | 1398 | 4139164 |