Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOTCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200001.1 | NP_001186930.1 | 8 | 1643 | ||
NM_024408.3 | NP_077719.2 | 8 | 1643 | |||
XM_005270901.3 | XP_005270958.1 | 9 | 3957 | |||
XM_011541519.2 | XP_011539821.1 | 8 | 6751 | |||
XM_011541520.2 | XP_011539822.1 | 8 | 3661 | |||
XM_017001372.1 | XP_016856861.1 | 8 | 1313 | |||
XM_017001373.1 | XP_016856862.1 | 8 | 1292 | |||
GenBank mRNA | AF308601.1 | - | 8 | 1358 | ||
AF315356.1 | - | 8 | 1358 | |||
AK307843.1 | - | 7 | 1484 | |||
BC071562.1 | - | 8 | 1592 | 4822955 | ||
GQ231534.1 | - | 8 | 1505 |