Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018089.2 | NP_001018099.1 | 4 | 415 | ||
NM_001276385.1 | NP_001263314.1 | 4 | 575 | |||
NM_024611.5 | NP_078887.2 | 4 | 575 | |||
XM_005254661.1 | XP_005254718.1 | 5 | 574 | |||
XM_005254662.1 | XP_005254719.1 | 5 | 437 | |||
XM_011522009.1 | XP_011520311.1 | 4 | 449 | |||
XM_011522010.1 | XP_011520312.1 | 4 | 318 | |||
XM_011522011.2 | XP_011520313.1 | 3 | 232 | |||
XM_017022569.1 | XP_016878058.1 | 5 | 611 | |||
GenBank mRNA | AF502591.1 | - | 4 | 518 | ||
AK055752.1 | - | 4 | 518 | |||
AK095389.1 | - | 4 | 399 | |||
AL832046.1 | - | 4 | 542 | |||
AY358546.1 | - | 4 | 536 | |||
BC020918.1 | - | 4 | 779 | |||
BC140894.1 | - | 4 | 494 | 9021762 | ||
BC151844.1 | - | 4 | 489 | |||
BX647873.1 | - | 4 | 401 | |||
CN342776.1 | - | 4 | 418 | |||
CR749852.1 | - | 4 | 368 |