Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018089.2 | NP_001018099.1 | 7 | 827 | ||
NM_001276385.1 | NP_001263314.1 | 7 | 987 | |||
NM_024611.5 | NP_078887.2 | 7 | 987 | |||
XM_005254661.1 | XP_005254718.1 | 8 | 986 | |||
XM_005254662.1 | XP_005254719.1 | 8 | 849 | |||
XM_011522009.1 | XP_011520311.1 | 7 | 861 | |||
XM_011522010.1 | XP_011520312.1 | 7 | 730 | |||
XM_011522011.2 | XP_011520313.1 | 6 | 644 | |||
XM_017022569.1 | XP_016878058.1 | 8 | 1023 | |||
GenBank mRNA | AF502591.1 | - | 7 | 930 | ||
AK055752.1 | - | 7 | 930 | |||
AK095389.1 | - | 7 | 811 | |||
AL832046.1 | - | 7 | 954 | |||
AY358546.1 | - | 7 | 948 | |||
BC020918.1 | - | 7 | 1191 | |||
BC140894.1 | - | 7 | 906 | 9021762 | ||
BC151844.1 | - | 7 | 901 | |||
BX647873.1 | - | 7 | 813 | |||
CR749852.1 | - | 7 | 780 |