Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018089.2 | NP_001018099.1 | 10 | 1754 | ||
NM_024611.5 | NP_078887.2 | 10 | 1914 | |||
XM_005254661.1 | XP_005254718.1 | 11 | 1913 | |||
XM_005254662.1 | XP_005254719.1 | 11 | 1776 | |||
XM_011522009.1 | XP_011520311.1 | 10 | 1788 | |||
XM_011522010.1 | XP_011520312.1 | 10 | 1657 | |||
XM_011522011.2 | XP_011520313.1 | 9 | 1571 | |||
XM_017022569.1 | XP_016878058.1 | 11 | 1950 | |||
GenBank mRNA | AF502591.1 | - | 10 | 1857 | ||
AK055752.1 | - | 10 | 1857 | |||
AK095389.1 | - | 10 | 1738 | |||
AK127892.1 | - | 3 | 739 | |||
AL832046.1 | - | 10 | 1881 | |||
AL833318.1 | - | 2 | 720 | |||
AY358546.1 | - | 10 | 1875 | |||
BC140894.1 | - | 10 | 1833 | 9021762 | ||
BX647873.1 | - | 10 | 1740 | |||
CR749852.1 | - | 10 | 1707 |