Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYBRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127383.1 | NP_001120855.1 | 3 | 2839 | ||
NM_001256909.1 | NP_001243838.1 | 4 | 2710 | |||
NM_024843.3 | NP_079119.3 | 4 | 3048 | |||
GenBank mRNA | AK026654.1 | - | 1 | 933 | ||
AK128495.1 | - | 3 | 2717 | |||
AL136693.1 | - | 4 | 2931 | |||
AL834483.1 | - | 1 | 2210 |