Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYBRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127383.1 | NP_001120855.1 | 3 | 3681 | ||
NM_001256909.1 | NP_001243838.1 | 4 | 3552 | |||
NM_024843.3 | NP_079119.3 | 4 | 3890 | |||
GenBank mRNA | AK026654.1 | - | 1 | 1775 | ||
AL136693.1 | - | 4 | 3773 | |||
AL834483.1 | - | 1 | 3052 |