Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROSER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025138.4 | NP_079414.3 | 11 | 3009 | ||
XM_005266545.3 | XP_005266602.1 | 10 | 2982 | |||
XM_011535239.2 | XP_011533541.1 | 10 | 2942 | |||
GenBank mRNA | AB107354.1 | - | 11 | 3009 | ||
AK022723.1 | - | 5 | 1685 | |||
AK074360.1 | - | 11 | 2981 | |||
AK299663.1 | - | 8 | 2005 | |||
AL833854.1 | - | 1 | 736 | |||
AL834435.1 | - | 1 | 225 | |||
BC053566.1 | - | 5 | 1688 | |||
BC064348.1 | - | 11 | 2264 | 4939198 | ||
BX647471.1 | - | 11 | 2431 |